Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylpentane, 99+%, ACROS Organics™

Click to view available options
Quantity:
50mL
250mL
1L
Packaging:
Glass bottle
96-14-0
C6H14
86.18
MFCD00009342
PFEOZHBOMNWTJB-UHFFFAOYSA-N
pentane, 3-methyl,unii-xd8o3ml76t,3-methyl-pentane,xd8o3ml76t,3-methylpentane, analytical standard,diethylmethylmethane,3-methylpentyl,3-methyl pentane,3-methylpentan-3-yl,acmc-209s6l
7282
3-methylpentane
CCC(C)CC
96-14-0
C6H14
86.18
MFCD00009342
PFEOZHBOMNWTJB-UHFFFAOYSA-N
pentane, 3-methyl,unii-xd8o3ml76t,3-methyl-pentane,xd8o3ml76t,3-methylpentane, analytical standard,diethylmethylmethane,3-methylpentyl,3-methyl pentane,3-methylpentan-3-yl,acmc-209s6l
7282
3-methylpentane
CCC(C)CC
Melting Point -118.0°C
Quantity 50mL
Density 0.6600g/mL
Boiling Point 64.0°C
Color Colorless
Refractive Index 1.3755 to 1.3775
Specific Gravity 0.66
Packaging Glass bottle
Flash Point −32°C
Infrared Spectrum Authentic
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula C2H5CH(CH3)C2H5
Beilstein 01, 149
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, slightly soluble in ether
Formula Weight 86.18
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material 3-Methylpentane, 99+%
Show More Show Less

GHS H Statement
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap

GHS Signal Word: Danger

EINECSNumber : 202-481-4

TSCA : TSCA