Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methylpentane, 99+%

Catalog No. p-166553
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle

CAS: 96-14-0 | C6H14 | 86.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-14-0
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD00009342
InChI Key PFEOZHBOMNWTJB-UHFFFAOYSA-N
Synonym pentane, 3-methyl, unii-xd8o3ml76t, 3-methyl-pentane, xd8o3ml76t, 3-methylpentane, analytical standard, diethylmethylmethane, 3-methylpentyl, 3-methyl pentane, 3-methylpentan-3-yl, acmc-209s6l
PubChem CID 7282
IUPAC Name 3-methylpentane
SMILES CCC(C)CC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -118.0°C
Color Colorless
Density 0.6600g/mL
Boiling Point 64.0°C
Flash Point −32°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.3755 to 1.3775
Linear Formula C2H5CH(CH3)C2H5
Quantity 50 mL
Beilstein 01, 149
Specific Gravity 0.66
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, slightly soluble in ether
Formula Weight 86.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 3-Methylpentane, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap
EINECSNumber 202-481-4
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.