Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Methylthio)propionaldehyde, 97%

Catalog No. p-7022144
Click to view available options
Quantity:
50 g
250 g

CAS: 3268-49-3 | C4H8OS | 104.167 g/mol

3-(Methylthio)propionaldehyde, is used as food flavour ,for making Baked food, spices, soft drinks, candy. It is also used as an intermediate. It has wide applications in organic synthesis as solvents as well as reagents. They are used in the synthesis of specific compound classes include agricultural chemical, property-enhancing additives, pharmacological drugs, chemical resistant polymers, detergents, and rubber antioxidants. Some examples of aliphatic and aromatic thioethers found in flavouring agents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Methylthio)propionaldehyde, is used as food flavour ,for making Baked food, spices, soft drinks, candy. It is also used as an intermediate. It has wide applications in organic synthesis as solvents as well as reagents. They are used in the synthesis of specific compound classes include agricultural chemical, property-enhancing additives, pharmacological drugs, chemical resistant polymers, detergents, and rubber antioxidants. Some examples of aliphatic and aromatic thioethers found in flavouring agents

Solubility
It is readily soluble in alcohol solvents such as ethanol, propylene and glycol oil. It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Air sensitive. Handle and store under inert gas. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3268-49-3
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.167
MDL Number MFCD00007022
InChI Key CLUWOWRTHNNBBU-UHFFFAOYSA-N
Synonym methional, 3-methylthio propionaldehyde, 3-methylthio propanal, propanal, 3-methylthio, 4-thiapentanal, 3-methylsulfanyl propanal, 3-methylmercapto propionaldehyde, 3-methylthiopropanal, propionaldehyde, 3-methylthio, 3-methylmercaptopropyl aldehyde
PubChem CID 18635
ChEBI CHEBI:49017
IUPAC Name 3-methylsulfanylpropanal
SMILES CSCCC=O

Specifications

Melting Point -58°C
Density 1.052
Boiling Point 165°C to 166°C
Flash Point 64°C (147°F)
Odor Strong
Refractive Index 1.4825
Linear Formula CH3S(CH2)2CHO
Quantity 250 g
UN Number UN2785
Beilstein 1739289
Sensitivity Air sensitive
Solubility Information It is readily soluble in alcohol solvents such as ethanol,propylene and glycol oil. It is insoluble in water.
Formula Weight 104.17
Percent Purity 98%
Chemical Name or Material 3-(Methylthio)propionaldehyde
Show More Show Less
Hazard Category H227-H302+H332-H311-H315-H317-H318
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H317-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 4-THIAPENTANAL
EINECSNumber 221-882-5
RTECSNumber UE2285000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.