Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(N,N-Dimethylpalmitylammonio)propanesulfonate, 98%

Catalog No. p-7077735
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 2281-11-0 | C21H45NO3S | 391.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2281-11-0
Molecular Formula C21H45NO3S
Molecular Weight (g/mol) 391.66
MDL Number MFCD00036911
InChI Key TUBRCQBRKJXJEA-UHFFFAOYSA-N
Synonym 3-n,n-dimethylpalmitylammonio propanesulfonate, zwittergent 3-16, 3-hexadecyldimethylammonio propane-1-sulfonate, palmityl sulfobetaine, n-hexadecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-hexadecyl dimethyl ammonio propane-1-sulfonate, 3-palmityldimethylammonio propanesulfonate, 3-hexadecyldimethylammonio propanesulfonate, 3-n,n-dimethylhexadecylammonio propanesulfonate, n-hexadecy-n,n-dimethyl-3-ammonio-1-propanesulfonate
PubChem CID 92176
IUPAC Name 3-[hexadecyl(dimethyl)azaniumyl]propane-1-sulfonate
SMILES CCCCCCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

Specifications

Elemental Analysis 3.5 to 3.7% (N, anhydrous) 63.1 to 65.7% (C, anhydrous)
Loss on Drying 2% max.
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information Solubility in water: 100g/L.
Formula Weight 391.65
Percent Purity 98%
Chemical Name or Material 3-(N,N-Dimethylpalmitylammonio)propanesulfonate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 218-918-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.