Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-n-Octylthiophene, 97%

Catalog No. p-7052715
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 65016-62-8 | C12H20S | 196.352 g/mol

3-Octylthiophene is used as a starting material in the synthesis of the following 2,5-dibromo-3-octylthiophene, poly(3-butylthiophene)-b-poly(3-octylthiophene), a diblock copoly(3-alkylthiophene), regioregular poly(3-octylthiophene).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Octylthiophene is used as a starting material in the synthesis of the following 2,5-dibromo-3-octylthiophene, poly(3-butylthiophene)-b-poly(3-octylthiophene), a diblock copoly(3-alkylthiophene), regioregular poly(3-octylthiophene).

Solubility
Soluble in organic solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65016-62-8
Molecular Formula C12H20S
Molecular Weight (g/mol) 196.352
MDL Number MFCD00085281
InChI Key WQYWXQCOYRZFAV-UHFFFAOYSA-N
Synonym 3-n-octylthiophene, thiophene, 3-octyl, acmc-1bfcd, 3-octylthiophene, ksc491i4b, bidd:gt0691
PubChem CID 566852
IUPAC Name 3-octylthiophene
SMILES CCCCCCCCC1=CSC=C1

Specifications

Color Colorless
Density 0.92 g/mL
Boiling Point 106°C to 107°C (3 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.492
Quantity 5 g
Solubility Information Soluble in organic solvents.
Formula Weight 196.35
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-n-Octylthiophene
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.