Learn More
3-Nitrophenol, 98+%
Description
3-Nitrophenol is used as indicator in 0.3% Soln in 50% alcohol ph: 6.8 Colorless, 8.6 Yellow. It is used in the synthesis of RAGE inhibitors via the etherification of phenols. Also used in the synthesis of kinase inhibitors against epidermal growth factor receptor T790M mutation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Nitrophenol is used as indicator in 0.3% Soln in 50% alcohol ph: 6.8 Colorless, 8.6 Yellow. It is used in the synthesis of RAGE inhibitors via the etherification of phenols. Also used in the synthesis of kinase inhibitors against epidermal growth factor receptor T790M mutation.
Solubility
Soluble in water (30 mg/ml), ether, alcohol, acetone, and ethanol (700 mg/ml).
Notes
Store in a tightly closed container. Keep from contact with oxidizing materials. Store in a cool, dry, well-ventilated area away from incompatible substances.
Specifications
Specifications
| Melting Point | 93°C to 98°C |
| Density | 1.49 |
| Boiling Point | 194°C (70 mmHg) |
| Flash Point | >100°C (212°F) |
| Quantity | 10 g |
| UN Number | UN1663 |
| Beilstein | 1907946 |
| Merck Index | 14,6619 |
| Solubility Information | Soluble in water (30mg/ml),ether,alcohol,acetone,and ethanol (700mg/ml). |
| Formula Weight | 139.11 |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H318-H373-H500 |
| Hazard Statement | GHS H Statement H318-H373-H302-H312-H335-H315 Causes serious eye damage. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful in contact with skin. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | P260-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P314-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROPHENOLS |
| EINECSNumber | 209-073-5 |
| RTECSNumber | SM1925000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.