Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Octylthiophene, 97%, Thermo Scientific Chemicals

Catalog No. p-176792
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65016-62-8
MDL Number MFCD00085281
InChI Key WQYWXQCOYRZFAV-UHFFFAOYSA-N
Synonym 3-n-octylthiophene, thiophene, 3-octyl, acmc-1bfcd, 3-octylthiophene, ksc491i4b, bidd:gt0691
PubChem CID 566852
IUPAC Name 3-octylthiophene
SMILES CCCCCCCCC1=CSC=C1

Specifications

CAS Min % 92.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 106°C to 107°C (3.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C12H20S
Refractive Index 1.4870 to 1.492
Quantity 10 g
Specific Gravity 0.91
Molecular Weight (g/mol) 196.36
Formula Weight 196.36
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Octylthiophene, 94%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.