Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-OHKYN Antibody (P3UI), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB100597IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100597IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100597IR Supplier Novus Biologicals Supplier No. NB100597IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

3-OHKYN Monoclonal specifically detects 3-OHKYN in All species, Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen 3-OHKYN
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone P3UI
Conjugate DyLight 755
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Alias 3039Hydroxykynurenine, 3-hydroxykynuenine modification
Host Species Mouse
Immunogen 3-OHKYN modified KLH
Molecular Weight of Antigen 66 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This is specific for 3-hydroxykynureinine (3OH-Kyn) modifications on proteins. It is likely to react with 3OH-Kyn modification on lysine, histidine and cysteine residues on proteins. It does not react with enzymes involved in the degradation of tryptopha
Target Species All species, Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.