Learn More
Corning™ 3D Clear Tissue Clearing Reagent
Catalog No. 10320106
Click to view available options
Quantity:
10 mL
30 mL
100 mL
Corning™'s 3D Clear tissue clearing technique is designed specifically to support imaging for 3D cell culture models and plate-based high-throughput processing. When paired with fluorescent labeling (e.g. fluorescent protein, immunofluorescence, chemical dyes) and high content confocal microscopy, Corning™'s 3D Clear Reagent allows for complete 3D cell culture model characterization and more accurate drug screening.
  • Rapid tissue clearing
  • Easy to Use
  • No Special Equipment Required
  • Compatible with IF, FP and other Fluorescent labels
  • Microplate and Automation Compatible
  • Reversible to Follow up with 2D H&E/IHC
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.