Learn More
3M™ Speedglas Helmet: Protection Plates
Designed for use with Speedglas Helmets
Supplier:  3M™ 7100009491
 View more versions of this product
Catalog No. 19-060-0430
Only null left
Add to Cart
Description
Description
3M offers a variety of replacement protection plates and accessories designed to protect the filter from spark and spatter.
  • Replacement inside protection plates for 3M™ Speedglas™ Filters 9002X and 9000X/XF/Xi
  • Speedglas 9000 standard outside protection plate
  • Replacement silver front panel (only) for 3M™ Speedglas™ FlexView filter assembly
  • Replacement face seal for 3M™ Speedglas™ Helmet 9000 HWR
  • Throat protector with Velcro Fasteners attaches to 3M™ Speedglas™ Helmets 9000 Series
  • Leather throat and side protectors for 3M™ Speedglas™ Fresh-air (FA) PA and SA (APC style helmet)
  • 3M Catalog# 04-0270-01 , 3M Catalog# 04-0280-00 , 3M Catalog# 18-0099-68
Sold separatelyFailure to use an outside protection plate with the filter voids the warranty if spark or spatter contaminates the glass. This option is standard on all Speedglas 9000 Series and Utility Welding Helmets. Outside protection plate made in USA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Utility Welding Helmets, Speedglas 9000 Series
Speedglas Helmet Accessories
04-0270-01
Helmet Outside Protection Plate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.