Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 3PO
GREENER_CHOICE

Catalog No. 512150
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

PFKFB3 inhibitor

3PO is an originally reported to inhibit PFKFB3 (IC50 = 25 μM). More recent reports failed to demonstrate activity in a PFKFB3 kinase assay. Reduces glycolytic flux and suppresses glucose uptake. Inhibits endothelial cell proliferation and causes G2/M cell cycle arrest in vitro. Attenuates vessel sprouting and tumor growth in vivo. Amplifies the antiangiogenic effect of VEGFR blockade.

Specifications

Quantity 50 mg
Product Type 3PO
Purity >99%
Molecular Weight (g/mol) 210.23
Inhibitors 3PO

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.