Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-1BB Ligand/TNFSF9 Antibody (AT113-2), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB152384 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB152384 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB152384 Supplier Novus Biologicals Supplier No. NBP311986AF532
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

4-1BB Ligand/TNFSF9 Monoclonal antibody specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence

Specifications

Antigen 4-1BB Ligand/TNFSF9
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone AT113-2
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9
Host Species Rabbit
Immunogen This antibody was raised by immunising LOU rats with soluble recombinant 4-1BB Ligand/TNFSF9:Fc fusion protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 8744
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.