Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-1BB Ligand/TNFSF9 Antibody (RM0067-3H19), HRP, Novus Biologicals™
SDP

Catalog No. NBP121514H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP121514H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP121514H Supplier Novus Biologicals Supplier No. NBP121514H
Add to Cart
Add to Cart

Rat Monoclonal Antibody

4-1BB Ligand/TNFSF9 Monoclonal specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen 4-1BB Ligand/TNFSF9
Applications Western Blot
Classification Monoclonal
Clone RM0067-3H19
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias 4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9
Gene Symbols TNFSF9
Host Species Rat
Immunogen Mouse recombinant 4-1BB Ligand EC-domain
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 8744
Test Specificity This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant 4-1BB Ligand Extracellular domain.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.