Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-1BB Ligand/TNFSF9 Antibody (TKS-1), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB016448 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB016448 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB016448 Supplier Novus Biologicals Supplier No. NBP281115V
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

4-1BB Ligand/TNFSF9 Monoclonal specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen 4-1BB Ligand/TNFSF9
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone TKS-1
Conjugate DyLight 405
Dilution Flow Cytometry, ELISA
Formulation 50mM Sodium Borate
Gene Alias 4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9
Gene Symbols TNFSF9
Host Species Rabbit
Immunogen Murine B-cell lymphoma 2PK-3 cells injected into SD rats.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 8744
Test Specificity This antibody binds to murine 4-1BB Ligand/TNFSF9 and competes with binding of 4-1BB.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.