Learn More
4-1BB Ligand/TNFSF9, Rabbit anti-Mouse, Clone: TKS-1, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | 4-1BB Ligand/TNFSF9 |
| Applications | Flow Cytometry, Blocking Assay |
| Classification | Monoclonal |
| Clone | TKS-1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry, Block/Neutralize |
| Formulation | PBS |
| Gene Alias | 4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9 |
| Gene Symbols | TNFSF9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.