Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-1BB/TNFRSF9/CD137 Antibody (2356B), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB372952 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB372952 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB372952 Supplier Novus Biologicals Supplier No. FAB8381L
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

4-1BB/TNFRSF9/CD137 Monoclonal specifically detects 4-1BB/TNFRSF9/CD137 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot.

Specifications

Antigen 4-1BB/TNFRSF9/CD137
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2356B
Conjugate DyLight 550
Dilution Western Blot, Simple Western, Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias 4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human 4-1BB/TNFRSF9/CD137, Leu24-His183, Accession # Q07011
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 3604
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.