Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-1BB/TNFRSF9/CD137 Antibody (JG1.6A), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N1064306PC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1064306PC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1064306PC5 Supplier Novus Biologicals Supplier No. NB10064306PECY55
Add to Cart
Add to Cart

Rat Monoclonal Antibody

4-1BB/TNFRSF9/CD137 Monoclonal specifically detects 4-1BB/TNFRSF9/CD137 in Human samples. It is validated for ELISA, Flow Cytometry.

Specifications

Antigen 4-1BB/TNFRSF9/CD137
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone JG1.6A
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias 4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9
Gene Symbols TNFRSF9
Host Species Rat
Immunogen Recombinant human CD137-Fc
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 3604.0
Test Specificity NB100-64306 recognizes human CD137, an 83kD type I integral membrane protein, otherwise known as 4-1BB. The CD137 molecule is primarily expressed on activated T-cells. It is a costimulatory member of the tumour necrosis receptor family and is reported to regulate T-cell activation and proliferation.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.