Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-1BB/TNFRSF9/CD137 Antibody (OTI5H5) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP271035 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271035 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271035 Supplier Novus Biologicals Supplier No. NBP271035
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

4-1BB/TNFRSF9/CD137 Monoclonal specifically detects 4-1BB/TNFRSF9/CD137 in Human samples. It is validated for Western Blot.

Specifications

Antigen 4-1BB/TNFRSF9/CD137
Applications Western Blot
Classification Monoclonal
Clone OTI5H5
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias 4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9
Gene Symbols TNFRSF9
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 24-186 of human TNFRSF9 (NP_001552) produced in E.coli.
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 3604
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.