Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-(1H-1,2,4-triazol-1-ylmethyl)benzoic acid, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
| CAS Min % | 96.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 97% |
| Packaging | Amber glass bottle |
| Quantity | 250 mg |
| Synonym | 4-1h-1,2,4-triazol-1-ylmethyl benzoic acid, 4-1h-1,2,4-triazol-1-yl methyl benzoic acid, 4-1,2,4-triazol-1-ylmethyl benzoic acid, 4-1,2,4 triazol-1-ylmethyl-benzoic acid, 4-1h-1,2,4-triazol-1-ylmethyl benzoic acid hydrochloride, 4-1,2,4-triazolylmethyl benzoic acid, 4-1h-1,2,4-triazole-1-ylmethyl benzoic acid, benzoic acid,4-1h-1,2,4-triazol-1-ylmethyl, benzoic acid, 4-1h-1,2,4-triazol-1-ylmethyl |
| Formula Weight | 203.2 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-(1H-1, 2, 4-triazol-1-ylmethyl)benzoic acid |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement: Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation. Flammable solid. |
| Precautionary Statement | GHS P Statement: Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep container tightly closed. Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction