Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2,5-Dioxotetrahydrofuran-3-yl)-1,2,3,4-tetrahydronaphthalene-1,2-dicarboxylic Anhydride 97.0+%, TCI America™
SDP

Catalog No. d21925g Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21925G 5 g
D219225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21925G Supplier TCI America Supplier No. D21925G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13912-65-7
Molecular Formula C16H12O6
Molecular Weight (g/mol) 300.266
MDL Number MFCD00030105
InChI Key JYCTWJFSRDBYJX-UHFFFAOYSA-N
Synonym 4-(2,5-Dioxotetrahydrofuran-3-yl)-tetralin-1,2-dicarboxylic Anhydride
PubChem CID 85657
IUPAC Name 5-(2,5-dioxooxolan-3-yl)-3a,4,5,9b-tetrahydrobenzo[e][2]benzofuran-1,3-dione
SMILES C1C(C2=CC=CC=C2C3C1C(=O)OC3=O)C4CC(=O)OC4=O

Specifications

Melting Point 204°C
Color White
Quantity 5 g
Formula Weight 300.27
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2,5-Dioxotetrahydrofuran-3-yl)-1,2,3,4-tetrahydronaphthalene-1,2-dicarboxylic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.