Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Aminoethyl)morpholine, 98+%

Catalog No. p-3756795
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 2038-03-1 | C6H14N2O | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2038-03-1
Molecular Formula C6H14N2O
Molecular Weight (g/mol) 130.19
MDL Number MFCD00006182
InChI Key RWIVICVCHVMHMU-UHFFFAOYSA-N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 24°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 205°C
Flash Point 175°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4745 to 1.4765
Quantity 5 g
Specific Gravity 0.99
Synonym 2',3',4',5',6'-pentamethylacetophenone, 1-pentamethylphenyl ethanone, 1-pentamethylphenyl ethan-1-one, 2,3,4,5,6-pentamethylacetophenone, 1-2,3,4,5,6-pentamethylphenyl ethanone, acmc-1cl24, 2,3,4,5,6-pentamethyl acetophenone, 1,2,3,4,5-pentamethyl-6-acetylbenzene, 1-acetyl-2,3,4,5,6-pentamethylbenzene, ethanone,1-2,3,4,5,6-pentamethylphenyl
Solubility Information Solubility in water: soluble
SMILES NCCN1CCOCC1
IUPAC Name 2-(morpholin-4-yl)ethan-1-amine
PubChem CID 74876
Formula Weight 130.19
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material 4-(2-Aminoethyl)morpholine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 218-011-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.