Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(2-Hydroxyethyl)morpholine, 99%

Click to view available options
Quantity:
100g
250g
500g
622-40-2
C6H13NO2
131.175
MFCD00006180
KKFDCBRMNNSAAW-UHFFFAOYSA-N
2-morpholinoethanol,4-2-hydroxyethyl morpholine,4-morpholineethanol,n-2-hydroxyethyl morpholine,2-4-morpholinyl ethanol,morpholine ethanol,morpholineethanol,hydroxyethylmorpholine,ethanol, 2-morpholino,beta-morpholinoethanol
61163
CHEBI:67144
2-morpholin-4-ylethanol
C1COCCN1CCO
622-40-2
C6H13NO2
131.175
MFCD00006180
KKFDCBRMNNSAAW-UHFFFAOYSA-N
2-morpholinoethanol,4-2-hydroxyethyl morpholine,4-morpholineethanol,n-2-hydroxyethyl morpholine,2-4-morpholinyl ethanol,morpholine ethanol,morpholineethanol,hydroxyethylmorpholine,ethanol, 2-morpholino,beta-morpholinoethanol
61163
CHEBI:67144
2-morpholin-4-ylethanol
C1COCCN1CCO
Applications
4-(2-Hydroxyethyl)morpholine is used as an organic chemical synthesis intermediate.

Solubility
Fully miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
Flash Point 99°C (210°F)
Boiling Point 226°C to 228°C
Quantity 100g
Melting Point 1°C to 2°C
pH 10.7
Beilstein 104375
Formula Weight 131.18
Percent Purity 99%
Chemical Name or Material 4-(2-Hydroxyethyl)morpholine

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : QE3510000

TSCA : TSCA