Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Oxo-1-pyrrolidinyl)benzene sulfonyl chloride, 97+%, Thermo Scientific™

Catalog No. JFD00486DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 112539-09-0
Molecular Formula C10H10ClNO3S
Molecular Weight (g/mol) 259.704
InChI Key OUAVTZYGHGCCCB-UHFFFAOYSA-N
Synonym 4-2-oxopyrrolidin-1-yl benzenesulfonyl chloride, 4-2-oxo-1-pyrrolidinyl benzenesulfonyl chloride, 4-2-oxo-pyrrolidin-1-yl-benzenesulfonylchloride, 4-2-oxopyrrolidin-1-yl benzene-1-sulfonyl chloride, 4-2-oxo-pyrrolidin-1-yl-benzenesulfonyl chloride, benzenesulfonylchloride, 4-2-oxo-1-pyrrolidinyl, 1-4-chlorosulfonyl phenyl pyrrolidin-2-one, pubchem5680, acmc-1bri4, benzenesulfonylchloride,4-2-oxo-1-pyrrolidinyl
PubChem CID 2794593
IUPAC Name 4-(2-oxopyrrolidin-1-yl)benzenesulfonyl chloride
SMILES C1CC(=O)N(C1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 259.71
Percent Purity 97%
Chemical Name or Material 4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.