Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4,4'-Diaminodiphenyl sulfone, 98%

Click to view available options
Quantity:
100g
500g
80-08-0
C12H12N2O2S
248.3
MFCD00007887
MQJKPEGWNLWLTK-UHFFFAOYSA-N
dapsone,diaphenylsulfone,4,4'-diaminodiphenyl sulfone,dapson,4,4'-sulfonyldianiline,diaphenylsulfon,sulfona,4-aminophenyl sulfone,avlosulfone,diphenasone
2955
CHEBI:4325
4-(4-aminophenyl)sulfonylaniline
C1=CC(=CC=C1N)S(=O)(=O)C2=CC=C(C=C2)N
80-08-0
C12H12N2O2S
248.3
MFCD00007887
MQJKPEGWNLWLTK-UHFFFAOYSA-N
dapsone,diaphenylsulfone,4,4'-diaminodiphenyl sulfone,dapson,4,4'-sulfonyldianiline,diaphenylsulfon,sulfona,4-aminophenyl sulfone,avlosulfone,diphenasone
2955
CHEBI:4325
4-(4-aminophenyl)sulfonylaniline
C1=CC(=CC=C1N)S(=O)(=O)C2=CC=C(C=C2)N
Solubility
Insoluble in water. Solubility in hot methanol is almost transparent. Soluble in DMSO, ethanol and acetone.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Odor Odorless
Quantity 500g
Melting Point 175°C to 178°C
Beilstein 788055
Merck Index 14,2822
Solubility Information Insoluble in water. Solubility in hot methanol is almost transparent. Soluble in DMSO,ethanol and acetone.
Formula Weight 248.3
Percent Purity 98%
Chemical Name or Material 4,4'-Diaminodiphenyl sulfone

GHS H Statement
H302
Harmful if swallowed.

P260-P264b-P270-P301+P312-P314-P330-P501c

H302-H373

EINECSNumber : 201-248-4

RTECSNumber : BY8925000

TSCA : Yes

Recommended Storage : Ambient temperatures