Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Iodobenzyl)morpholine, ≥97%, Thermo Scientific™

Catalog No. CC29311DE
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC29311DE 5 g
CC29311DA 1 g
CC29311EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC29311DE Supplier Thermo Scientific Chemicals Supplier No. CC29311DE
Add to Cart

Chemical Identifiers

CAS 299159-27-6
Molecular Formula C11H14INO
Molecular Weight (g/mol) 303.143
MDL Number MFCD04974051
InChI Key IYDFKIKPNVVXQU-UHFFFAOYSA-N
Synonym 4-4-iodobenzyl morpholine, 4-4-iodophenyl methyl morpholine, 4-4-iodo-benzyl-morpholine, 4-iodobenzyl morpholine, 4-morpholinomethyliodobenzene
PubChem CID 2795500
IUPAC Name 4-[(4-iodophenyl)methyl]morpholine
SMILES C1COCCN1CC2=CC=C(C=C2)I

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 5 g
Formula Weight 303.14
Percent Purity 95%
Chemical Name or Material 4-(4-Iodobenzyl)morpholine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.