Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium metatungstate, 2.82 density solution in water, stabilized

Catalog No. AC304102500
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle

CAS: 12141-67-2 | H2Na6O40W12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12141-67-2
Molecular Formula H2Na6O40W12
MDL Number MFCD00150663
Synonym Sodium polytungstate

Specifications

Color Green to Yellow
Linear Formula Na6[H2W12O40]
Density 2.8200g/mL
Solubility Information Solubility in water: >1000g/L water (20°C).
Formula Weight 2986.13
Percent Purity ≥67% (WO3)
Packaging Glass bottle
Specific Gravity 2.82
Quantity 250 mL
Physical Form Liquid
Chemical Name or Material Sodium metatungstate, 2.82 Density Solution in Water, Stabilized
Show More Show Less
missing translation for 'healthHazard1' Danger
missing translation for 'healthHazard2' GHS H Statement:
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
missing translation for 'healthHazard3' GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.