Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 4μ8C
GREENER_CHOICE

Catalog No. 447910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-791-0 10 mg
44-795-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-791-0 Supplier Tocris Bioscience™ Supplier No. 4479/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibits IRE1α endoribonuclease activity

Selective inhibitor of IRE1α ribonuclease (RNase) activity (IC50 = 60 nM). Blocks substrate access to the active site of IRE1α. Inactivates IRE1α-mediated mRNA degradation. Displays no effect on the IRE1α-related endoribonuclease RNase L.

Chemical Identifiers

CAS 14003-96-4
Molecular Formula C11H8O4

Specifications

Quantity 10 mg
Formula Weight 204.18
Purity >98%
Molecular Weight (g/mol) 204.18
Chemical Name or Material 4μ8C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.