Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Amino-2-methyl-5-pyrimidinemethanol 98.0+%, TCI America™
SDP

Catalog No. A27301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27301G 1 g
A2730200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27301G Supplier TCI America Supplier No. A27301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 73-67-6
Molecular Formula C6H9N3O
Molecular Weight (g/mol) 139.158
MDL Number MFCD00849239
InChI Key VUTBELPREDJDDH-UHFFFAOYSA-N
Synonym 4-Amino-5-hydroxymethyl-2-methylpyrimidine
PubChem CID 777
ChEBI CHEBI:16892
IUPAC Name (4-amino-2-methylpyrimidin-5-yl)methanol
SMILES CC1=NC=C(C(=N1)N)CO

Specifications

Melting Point 198°C
Color White-Yellow
Quantity 1 g
Formula Weight 139.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Amino-2-methyl-5-pyrimidinemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.