Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-amino Biphenyl DNA Antibody (4C11), Novus Biologicals™
SDP

Catalog No. NB100415SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100415SS 0.025 mL
NB100415 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100415SS Supplier Novus Biologicals Supplier No. NB100415SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

4-amino Biphenyl DNA Monoclonal specifically detects 4-amino Biphenyl DNA in All species, Mouse samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry.

Specifications

Antigen 4-amino Biphenyl DNA
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 4C11
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50 1:100
Formulation Ascites with 0.1% Sodium Azide
Gene Alias 4-AMBP DNA, 4-aminobiphenyl DNA, 4-amino-Biphenyl DNA
Host Species Mouse
Immunogen 4-amino Biphenyl DNA
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects DNA from all species.
Target Species All species, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.