Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobenzo-18-crown-6 sesquihydrate hydrochloride, 99%, Thermo Scientific™

Catalog No. AC277462500
Click to view available options
Quantity:
250 mg
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205504-06-9
Molecular Formula 1·5 H2O
Molecular Weight (g/mol) 390.85
MDL Number MFCD00149387
InChI Key WLKFVHSGZPPEKQ-UHFFFAOYSA-N
Synonym 4-aminobenzo-18-crown-6 sesquihydrate hydrochloride, 2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecin-18-amine hydrate hydrochloride
PubChem CID 53393546
IUPAC Name 2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(18),19,21-trien-20-amine;hydrate;hydrochloride
SMILES C1COCCOCCOC2=C(C=CC(=C2)N)OCCOCCO1.O.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 113°C to 117°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 250 mg
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol
Formula Weight 390.85
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Aminobenzo-18-crown-6 sesquihydrate hydrochloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.