Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobutyraldehyde dimethyl acetal, 98+%

Catalog No. aab2107206
Click to view available options
Quantity:
5 g
25 g

CAS: 19060-15-2 | C6H16NO2 | 134.20 g/mol

4-Aminobutyraldehyde dimethyl acetal acts as an intermediate in organic synthesis. It is used in the preparation of pineal hormone melatonin by reaction with 4-methoxyphenylhydrazine hydrochloride and acetic anhydride. Further, it is also used in the preparation of ficuseptine, juliprosine, and juliprosopine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobutyraldehyde dimethyl acetal acts as an intermediate in organic synthesis. It is used in the preparation of pineal hormone melatonin by reaction with 4-methoxyphenylhydrazine hydrochloride and acetic anhydride. Further, it is also used in the preparation of ficuseptine, juliprosine, and juliprosopine.

Solubility
Miscible with benzene.

Notes
Air sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19060-15-2
Molecular Formula C6H16NO2
Molecular Weight (g/mol) 134.20
MDL Number MFCD00189373
InChI Key TYVAXMOICMBSMT-UHFFFAOYSA-O
Synonym 4-aminobutyraldehyde dimethyl acetal, 4,4-dimethoxybutylamine, 4,4-dimethoxy-1-butanamine, 4-aminobutyraldehyde dimethylacetal, acmc-1bqj6, 4-aminobutyraldehyde dimethyl, 1-amino-4,4-dimethoxybutane, cs-waa0041, tyvaxmoicmbsmt-uhfffaoysa, 4-aminobutyraldehydedimethylacetal
PubChem CID 5463803
SMILES COC(CCC[NH3+])OC

Specifications

Boiling Point 85°C (22 mmHg)
Flash Point 59°C (138°F)
Refractive Index 1.43
Linear Formula H2N(CH2)3CH(OCH3)2
Quantity 5 g
UN Number UN2734
Sensitivity Air Sensitive
Solubility Information Miscible with benzene.
IUPAC Name 4,4-dimethoxybutan-1-aminium
Formula Weight 133.19
Percent Purity ≥98%
Chemical Name or Material 4-Aminobutyraldehyde dimethyl acetal
Show More Show Less
Hazard Category H226-H302-H314-H334-H335
Hazard Statement GHS H Statement
H314-H226-H302-H317
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 407-690-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.