Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Aminobutyric acid, 97%

Catalog No. p-7081138
Click to view available options
Quantity:
100 g
500 g
2500 g

CAS: 56-12-2 | C4H9NO2 | 103.121 g/mol

In the production of polymers, food additives, and pharmaceuticals. Acrylic acid and methacrylic acids are well known precursors of polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In the production of polymers, food additives, and pharmaceuticals. Acrylic acid and methacrylic acids are well known precursors of polymers.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-12-2
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00008226
InChI Key BTCSSZJGUNDROE-UHFFFAOYSA-N
Synonym 4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino
PubChem CID 119
ChEBI CHEBI:16865
IUPAC Name 4-aminobutanoic acid
SMILES C(CC(=O)O)CN

Specifications

Melting Point ∼195°C (decomposition)
Linear Formula H2N(CH2)3CO2H
Beilstein 906818
Merck Index 14,430
Solubility Information Soluble in water.
Quantity 100 g
Odor Odorless
Formula Weight 103.12
Percent Purity 97%
Chemical Name or Material 4-Aminobutyric acid
EINECSNumber 200-258-6
RTECSNumber ES6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.