Learn More
4-Aminobutyric acid, 97%
Description
In the production of polymers, food additives, and pharmaceuticals. Acrylic acid and methacrylic acids are well known precursors of polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
In the production of polymers, food additives, and pharmaceuticals. Acrylic acid and methacrylic acids are well known precursors of polymers.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | ∼195°C (decomposition) |
| Linear Formula | H2N(CH2)3CO2H |
| Beilstein | 906818 |
| Merck Index | 14,430 |
| Solubility Information | Soluble in water. |
| Quantity | 2500 g |
| Odor | Odorless |
| Formula Weight | 103.12 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Aminobutyric acid |
Safety and Handling
| EINECSNumber | 200-258-6 |
| RTECSNumber | ES6300000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.