Learn More
4-Aminobutyric acid, 99+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• 4-Aminobutyric acid is a γ-amino acid and monocarboxylic acid derived from butyric acid
• It is an amino acid that shows inhibitory activities against neurotransmitters, including GABA, and acts by binding GABA-A receptor and inhibiting the activity of signal-receiving neurons
• This compound, converted from neurotransmitter glutamate, is essential to regulate neuronal excitability
• It binds to GABA-A and GABA-B receptors, causing ion channel opening, hyperpolarization, and eventually inhibition of neurotransmission
Applications
• 4-Aminobutyric acid has been used in the preparation of GABA–BSA (bovine serum albumin)–GA (glutaraldehyde) conjugate, which is used as a control for immunohistochemical experiments
• It is used, in vivo, to detect GABA neurotransmitter by magnetic resonance spectroscopy
• This compound increases the production of short-chain fatty acids and decreases pH values in mouse colon
• Some in vivo experiments, using rats, mice, and pigs, demonstrated that 4-Aminobutyric acid can modulate intestinal fluid secretion, blood flow and ulceration
Specifications
Specifications
| Melting Point | 197°C to 204°C |
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Color | White |
| Infrared Spectrum | Authentic |
| Loss on Drying | 0.5% max. |
| Assay Percent Range | 99+% |
| Packaging | Plastic Bottle |
| Linear Formula | H2N(CH2)3CO2H |
| Beilstein | 04,413 |
| Show More |
Safety and Handling
| EINECSNumber | 200-258-6 |
| RTECSNumber | ES6300000 |
| TSCA | TSCA |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.