Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Azabenzimidazole, 98%

Catalog No. AAB2505106
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 273-21-2 | C6H5N3 | 119.13 g/mol

4-Azabenzimidazole is used in the preparation of imidazo[4,5-b]pyridine 4-oxide. It is also used as a reagent to prepare benzoylmethyl-substituted azoles which are inhibitors of nitric oxide synthase with antioxidant properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Azabenzimidazole is used in the preparation of imidazo[4,5-b]pyridine 4-oxide. It is also used as a reagent to prepare benzoylmethyl-substituted azoles which are inhibitors of nitric oxide synthase with antioxidant properties.

Solubility
Soluble in dimethylformamide.

Notes
Incompatible with strong oxidizing agent and strong acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 273-21-2
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.13
MDL Number MFCD00005579,MFCD01646138
InChI Key GAMYYCRTACQSBR-UHFFFAOYSA-N
Synonym 3h-imidazo 4,5-b pyridine, 4-azabenzimidazole, 1h-imidazo 4,5-b pyridine, 1-deazapurine, 3,4-diazaindole, 7-azabenzimidazole, imidazopyridine, 4h-imidazo 4,5-b pyridine, pyrido 2,3-d imidazole, azabenzimidazole
PubChem CID 67504
ChEBI CHEBI:59952
IUPAC Name 1H-imidazo[4,5-b]pyridine
SMILES N1C=NC2=NC=CC=C12

Specifications

Melting Point 145°C to 150°C
Boiling Point 140°C to 150°C (0.1 mmHg)
Quantity 5 g
Beilstein 2600
Solubility Information Soluble in dimethylformamide.
Formula Weight 119.13
Percent Purity 99%
Chemical Name or Material 4-Azabenzimidazole
EINECSNumber 205-987-3
RTECSNumber NJ5108000
TSCA No

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.