Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Benzyloxy-3-methoxybenzyl alcohol, 97%, Thermo Scientific™

Catalog No. p-65515
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33693-48-0
Molecular Formula C15H16O3
Molecular Weight (g/mol) 244.29
MDL Number MFCD00004640
InChI Key PDBXFVPMVYQICB-UHFFFAOYSA-N
Synonym 4-benzyloxy-3-methoxybenzyl alcohol, 4-benzyloxy-3-methoxyphenyl methanol, 4-benzyloxy-3-methoxybenzylalcohol, benzenemethanol, 3-methoxy-4-phenylmethoxy, acmc-20amos, o-benzylvanillyl alcohol, 4-o-benzyl vanillyl alcohol, vanillyl alcohol benzyl ether
PubChem CID 98832
IUPAC Name (3-methoxy-4-phenylmethoxyphenyl)methanol
SMILES COC1=C(C=CC(=C1)CO)OCC2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 70°C to 73°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5CH2OC6H3(OCH3)CH2OH
Quantity 1 g
Formula Weight 244.29
Percent Purity ≥96%
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxy-3-methoxybenzyl alcohol, 97%
Show More Show Less
EINECSNumber 251-638-3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.