Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Biphenyl isocyanate, 97%

Catalog No. AC438500010
Click to view available options
Quantity:
1 g
5 g

CAS: 92-95-5 | C13H9NO | 195.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-95-5
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.22
MDL Number MFCD00037089
InChI Key WIRPZDICFIIBRF-UHFFFAOYSA-N
Synonym 4-biphenylyl isocyanate, p-xenylcarbimide, 4-biphenyl isocyanate, 4-isocyanatobiphenyl, 4-phenylphenyl isocyanate, unii-b0zsp4upq4, 4-biphenylylisocyanate, b0zsp4upq4, p-diphenyl isocyanate, p-xenylcarbimide mi
PubChem CID 3612319
IUPAC Name 1-isocyanato-4-phenylbenzene
SMILES O=C=NC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 60.0°C to 61.0°C
Color Brown to Orange
Boiling Point 110.0°C to 112.0°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15,10271
Formula Weight 195.22
Percent Purity 97%
Physical Form Solution
Chemical Name or Material 4-Biphenyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.