Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Boc-amino)-N-Fmoc-L-phenylalanine, 95%

Catalog No. p-7034349
Click to view available options
Quantity:
250 mg
1 g

Catalog No. AAH52031MD

Supplier: Thermo Scientific Chemicals H52031MD

Only null left
Add to Cart

CAS: 174132-31-1 | C29H30N2O6 | 502.567 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174132-31-1
Molecular Formula C29H30N2O6
Molecular Weight (g/mol) 502.567
MDL Number MFCD00237007
InChI Key KVUAOWDVYMUKPE-VWLOTQADSA-N
Synonym s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxycarbonyl amino phenyl propanoic acid, fmoc-l-4-aminophe boc, fmoc-phe 4-nhboc-oh, fmoc-l-phe 4-nhboc-oh, 2s-3-4-tert-butoxycarbonyl amino phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-p-amino-phe boc-oh, ambotzfaa1741, fmoc-4-boc-amino-l-phenylalanine, s-fmoc-4-boc-amino phenylalanine, n-9h-fluorene-9-ylmethoxycarbonyl-4-tert-butoxycarbonylamino-l-phenylalanine
PubChem CID 2756117
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]propanoic acid
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 250 mg
Formula Weight 502.57
Percent Purity 95%
Chemical Name or Material 4-(Boc-amino)-N-Fmoc-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.