Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytarabine, 98%

Catalog No. AC449560050
Click to view available options
Quantity:
100 mg
1 g
5 g
Packaging:
Glass bottle

Cytarabine, CAS # 147-94-4, is an inhibitor of DNA synthesis that is used as an anticancer and antiviral agent. | CAS: 147-94-4 | C9H13N3O5 | 243.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Cytarabine is a nucleoside analog that acts as an antileukemic agent
  • Cytarabine becomes cytotoxic following conversion to its triphosphorylated form, which can be incorporated into DNA primers by DNA polymerase alpha
  • Can induce S phase cell cycle arrest and apoptosis in leukemic and neuronal cells

Applications

  • Has been used to study chemotherapeutic regimens in laboratory studies of leukemia
  • Useful for laboratory studies of cytarabine resistance mechanisms
  • At nanomolar concentrations, cytarabine has been seen to induce mitochondrial apoptosis in leukemic cells through upregulation of multiple Bcl-2 family members
  • It can induce neuronal cell death via inducing γ-H2AX phosphorylation in histones, in the absence of double-stranded DNA breaks
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147-94-4
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.22
InChI Key UHDGCWIWMRVCDJ-CCXZUQQUSA-N
Synonym cytarabine, cytosine arabinoside, ara-c, arabinocytidine, cytosar, arabinosylcytosine, aracytidine, cytarabin, cytarabinoside, tarabine
PubChem CID 6253
ChEBI CHEBI:28680
IUPAC Name 4-amino-1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)O

Specifications

Melting Point 220.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 152781
Solubility Information Solubility in water: soluble. Other solubilities: soluble in dmso, ethanol
Formula Weight 243.22
Percent Purity 98%
Chemical Name or Material Cytarabine
Hazard Category Serious eye damage/eye irritation (category 2), Germ cell mutagenicity (category 2), Reproductive toxicity (category 2), Skin sensitisation (category 1)
Hazard Statement Causes serious eye irritation, Suspected of causing genetic defects, Suspected of damaging fertility or the unborn child, May cause an allergic skin reaction
EINECSNumber 205-705-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.