Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate, 97%

Catalog No. AC367502500
Click to view available options
Quantity:
250 mg
500 mg
1 g
5 g
Packaging:
Glass bottle

Catalog No. AC367502500

Supplier: Thermo Scientific Chemicals 367502500

Only null left
Add to Cart

CAS: 98014-14-3 | C20H16Cl2N4Ru | 484.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98014-14-3
Molecular Formula C20H16Cl2N4Ru
Molecular Weight (g/mol) 484.35
MDL Number MFCD00150005
InChI Key MGAJEYXQYJBLQL-UHFFFAOYSA-L
Synonym cis-bis 2,2'-bipyridine dichlororuthenium ii hydrate, bis bipyridyl dichlororuthenium hydrate, 2,2'-bipyridine-dichlororuthenium-water 2/1/1, cis-dichlorobis 2,2'-bipyridine ruthenium ii, cis-bis 2,2'-bipyridine dichlororuthenium ii monohydrate, cis-bis 2,2-bipyridine dichlororuthenium ii hydrate, cis-bis-2,2'-bipyridine ruthenium ii chloride,hydrate, cis-bis 2,2 inverted exclamation marka-bipyridine dichlororuthenium ii hydrate
PubChem CID 16211844
SMILES Cl[Ru]Cl.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1

Specifications

Color Black to Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Based on Carbon)
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: soluble
IUPAC Name dichlororuthenium;2-pyridin-2-ylpyridine;hydrate
Formula Weight 484.35
Percent Purity 97%
Physical Form Powder
Chemical Name or Material cis-Bis(2, 2'-bipyridine)dichlororuthenium(II) hydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.