Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-(methylthio)pyrimidine 98.0+%, TCI America™
SDP

Catalog No. C308725G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Catalog No. C308725G

Supplier: TCI America C308725G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 49844-90-8
Molecular Formula C5H5ClN2S
Molecular Weight (g/mol) 160.62
MDL Number MFCD00006083
InChI Key DFOHHQRGDOQMKG-UHFFFAOYSA-N
Synonym 4-chloro-2-methylthio pyrimidine, 4-chloro-2-methylthiopyrimidine, 4-chloro-2-methylsulfanyl pyrimidine, 4-chloro-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-2-methylthio, 4-chloro-2-methylmercapto pyrimidine, zlchem 575, pubchem15403, pubchem15764, acmc-1asbn
PubChem CID 97113
IUPAC Name 4-chloro-2-(methylsulfanyl)pyrimidine
SMILES CSC1=NC=CC(Cl)=N1

Specifications

Boiling Point 112°C
Quantity 25 g
UN Number 1760
Formula Weight 160.62
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloro-2-(methylthio)pyrimidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.