Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-methylthiopyrimidine, 97%

Catalog No. AC163330100
Click to view available options
Quantity:
10 g
50 g

CAS: 49844-90-8 | C5H5ClN2S | 160.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49844-90-8
Molecular Formula C5H5ClN2S
Molecular Weight (g/mol) 160.62
MDL Number MFCD00006083
InChI Key DFOHHQRGDOQMKG-UHFFFAOYSA-N
Synonym 4-chloro-2-methylthio pyrimidine, 4-chloro-2-methylthiopyrimidine, 4-chloro-2-methylsulfanyl pyrimidine, 4-chloro-2-methylsulfanyl-pyrimidine, pyrimidine, 4-chloro-2-methylthio, 4-chloro-2-methylmercapto pyrimidine, zlchem 575, pubchem15403, pubchem15764, acmc-1asbn
PubChem CID 97113
IUPAC Name 4-chloro-2-methylsulfanylpyrimidine
SMILES CSC1=NC=CC(Cl)=N1

Specifications

Melting Point -2.0°C
Density 1.3810g/mL
Boiling Point 139.0°C to 140.0°C (36.0 mmHg)
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6004
Quantity 10 g
Beilstein 23, 371
Specific Gravity 1.381
Formula Weight 160.63
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methylthiopyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 256-500-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.