Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-2-nitrotoluene, 99%

Catalog No. p-7026979
Click to view available options
Quantity:
100 g
500 g

CAS: 89-59-8 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-59-8
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007215
InChI Key SQFLFRQWPBEDHM-UHFFFAOYSA-N
Synonym 4-chloro-2-nitrotoluene, 2-nitro-4-chlorotoluene, benzene, 4-chloro-1-methyl-2-nitro, 4,2-chloronitrotoluene, p-chloro-o-nitrotoluene, toluene, 4-chloro-2-nitro, 4-chloro-1-methyl-2-nitro-benzene, ccris 3116, 4-chlor-2-nitrotoluol, pubchem18919
PubChem CID 6975
IUPAC Name 4-chloro-1-methyl-2-nitrobenzene
SMILES CC1=CC=C(Cl)C=C1[N+]([O-])=O

Specifications

Melting Point 34°C to 38°C
Boiling Point 242°C
Flash Point 120°C (248°F)
Quantity 100 g
UN Number UN3457
Formula Weight 171.58
Percent Purity 99%
Chemical Name or Material 4-Chloro-2-nitrotoluene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLORONITROTOLUENES, SOLID
EINECSNumber 201-921-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.