Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-methoxy-2-methylpyridine N-Oxide 98.0+%, TCI America™
SDP

Catalog No. C240225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C240225G 25 g
C24025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C240225G Supplier TCI America Supplier No. C240225G

Chemical Identifiers

CAS 122307-41-9
Molecular Formula C7H8ClNO2
Molecular Weight (g/mol) 173.60
MDL Number MFCD03093995
InChI Key TWXMQDRFBLSXFN-UHFFFAOYSA-N
PubChem CID 5083929
IUPAC Name 4-chloro-3-methoxy-2-methylpyridin-1-ium-1-olate
SMILES COC1=C(Cl)C=C[N+]([O-])=C1C

Specifications

Melting Point 100°C
Quantity 25 g
Formula Weight 173.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-methoxy-2-methylpyridine N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.