Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate, 97%

Catalog No. p-7045279
Click to view available options
Quantity:
1 g
5 g

CAS: 23163-86-2 | C8H3ClF3NS | 237.624 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23163-86-2
Molecular Formula C8H3ClF3NS
Molecular Weight (g/mol) 237.624
MDL Number MFCD00041090
InChI Key AHFPRSSHNSGRCU-UHFFFAOYSA-N
Synonym 4-chloro-3-trifluoromethyl phenyl isothiocyanate, 1-chloro-4-isothiocyanato-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl phenylisothiocyanate, 4-chloro-3-trifluoromethylphenyl isothiocyanate, 1-chloro-4-isothiocyanato-2-trifluoromethyl-benzene, benzene, 1-chloro-4-isothiocyanato-2-trifluoromethyl, 4-chloro-3-trifluoromethyl benzenisothiocyanate, acmc-20amlf, pubchem5016, 4-chloro-3-trifluoromethylphenylisothiocyanate
PubChem CID 520026
IUPAC Name 1-chloro-4-isothiocyanato-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1N=C=S)C(F)(F)F)Cl

Specifications

Density 1.47
Boiling Point 248°C to 251°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.59
Quantity 1 g
UN Number UN2922
Beilstein 787371
Sensitivity Moisture sensitive
Formula Weight 237.63
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)phenyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.