Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophenylacetyl chloride, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
25026-34-0
C8H6Cl2O
189.04
MFCD00037111
UMQUIRYNOVNYPA-UHFFFAOYSA-N
4-chlorophenylacetyl chloride,p-chlorophenylacetyl chloride,2-4-chlorophenyl acetyl chloride,4-chlorobenzeneacetyl chloride,benzeneacetyl chloride, 4-chloro,4-chlorophenylacetylchloride,acmc-1cch1,4-chlorobenzeneacetylchloride,4-chlorobenzenacetyl chloride,p-chlorophenyl-acetyl chloride
90692
2-(4-chlorophenyl)acetyl chloride
C1=CC(=CC=C1CC(=O)Cl)Cl
25026-34-0
C8H6Cl2O
189.04
MFCD00037111
UMQUIRYNOVNYPA-UHFFFAOYSA-N
4-chlorophenylacetyl chloride,p-chlorophenylacetyl chloride,2-4-chlorophenyl acetyl chloride,4-chlorobenzeneacetyl chloride,benzeneacetyl chloride, 4-chloro,4-chlorophenylacetylchloride,acmc-1cch1,4-chlorobenzeneacetylchloride,4-chlorobenzenacetyl chloride,p-chlorophenyl-acetyl chloride
90692
2-(4-chlorophenyl)acetyl chloride
C1=CC(=CC=C1CC(=O)Cl)Cl
Specific Gravity 1.292
Refractive Index 1.5495 to 1.5565
Packaging Glass bottle
Density 1.2920g/mL
Quantity 5g
Infrared Spectrum Authentic
Color Brown to Yellow
Flash Point >110°C
Boiling Point 85.0°C (1.0 mmHg)
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Linear Formula ClC6H4CH2COCl
Solubility Information Solubility in water: reacts.
Formula Weight 189.04
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 4-Chlorophenylacetyl chloride, 98%
Show More Show Less

GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 246-571-1