Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-chlorotoluene, 98%
Click to view available options
Quantity:
25 mL
100 mL
1 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 6.0°C to 8.0°C |
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Density | 1.0700g/mL |
| Boiling Point | 162.0°C |
| Quantity | 25 mL |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Physical Form | Liquid |
| Refractive Index | 1.5190 to 1.5210 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Flammable liquid and vapor. May cause an allergic skin reaction. Harmful if inhaled. Toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. If skin |
| EINECSNumber | 203-397- |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction