Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Dimethylaminobenzylamine Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. D162025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D162025G 25 g
D16205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D162025G Supplier TCI America Supplier No. D162025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34403-52-6
Molecular Formula C9H15N2
Molecular Weight (g/mol) 151.23
MDL Number MFCD00012862
InChI Key PDJZOFLRRJQYBF-UHFFFAOYSA-O
PubChem CID 2734633
IUPAC Name [4-(dimethylamino)phenyl]methanaminium
SMILES CN(C)C1=CC=C(C[NH3+])C=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 223.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminobenzylamine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.