Learn More
4-Ethoxybenzyl alcohol, 98%
Description
It is used in the identification of sulfation sites of metabolites and prediction of the compounds’ biological effects and in chemical investigation of Wedelia calendulacea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used in the identification of sulfation sites of metabolites and prediction of the compounds’ biological effects and in chemical investigation of Wedelia calendulacea.
Solubility
Solubility in methanol is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 32°C to 34°C |
| Boiling Point | 272°C to 274°C |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.532 |
| Quantity | 50 g |
| Formula Weight | 152.19 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Ethoxybenzyl alcohol |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 228-283-8 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.