Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethoxymethylene-2-phenyloxazolin-5-one, 98+%

Catalog No. p-7080550
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAA1015503

Supplier: Thermo Scientific Chemicals A1015503

Only null left
Add to Cart

CAS: 15646-46-5 | C12H11NO3 | 217.224 g/mol

4-Ethoxymethylene-2-phenyloxazolin-5-one acts as sensitizing agent. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethoxymethylene-2-phenyloxazolin-5-one acts as sensitizing agent. It is also used as pharmaceutical intermediates.

Solubility
Soluble in methanol (50 mg/ml), and water (partly).

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15646-46-5
Molecular Formula C12H11NO3
Molecular Weight (g/mol) 217.224
MDL Number MFCD00003204
InChI Key SJHPCNCNNSSLPL-NTMALXAHSA-N
Synonym oxazolone, oxazolone sensitizing agent, 5 4h-oxazolone, 4-ethoxymethylene-2-phenyl, unii-78il0j7n72, 4-ethoxymethylene-2-phenyl-2-oxazolin-5-one, 4-ethoxymethylene-2-phenyloxazolin-5-one, 4-ethoxymethylene-2-phenyl-2-oxazoline-5-one, 4z-4-ethoxymethylidene-2-phenyl-1,3-oxazol-5-one, 2-oxazolin-5-one, 4-ethoxymethylene-2-phenyl, 4-ethoxymethylene-2-phenyloxazolone
PubChem CID 1712094
IUPAC Name (4Z)-4-(ethoxymethylidene)-2-phenyl-1,3-oxazol-5-one
SMILES CCOC=C1C(=O)OC(=N1)C2=CC=CC=C2

Specifications

Melting Point 93°C to 97°C
Quantity 1 g
Beilstein 163055
Solubility Information Soluble in methanol (50mg/ml),and water (partly).
Formula Weight 217.22
Percent Purity ≥98%
Chemical Name or Material 4-Ethoxymethylene-2-phenyloxazolin-5-one
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 239-713-9
RTECSNumber RQ5786250
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.